missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BMS CCR2 22
GREENER_CHOICE

Product Code. 18085912
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 18085912

Brand: Tocris Bioscience™ 3129/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity, potent CCR2 antagonist

High affinity CCR2 chemokine receptor antagonist (IC50 = 5.1 nM). Displays potent functional antagonism, IC50 values are 1 and 18 nM for chemotaxis and antagonism of calcium flux respectively. Selective over CCR3.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445479-97-0
Molecular Formula C28H34F3N5O4S

Specifications

Quantity 10 mg
Formula Weight 593.66
Purity >98%
Molecular Weight (g/mol) 593.666
Chemical Name or Material BMS CCR2 22
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.