missing translation for 'onlineSavingsMsg'
Learn More

BMS-911543, MedChemExpress

Product Code. 16498997
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498997 5 mg 5mg
16472517 50 mg 50mg
16440236 2 mg 2mg
16426706 100 mg 100mg
16406418 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498997 Supplier Medchem Express Supplier No. HY15270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-911543 is a selective JAK2 inhibitor, with IC50s of 1.1 nM, less selective at JAK1, JAK3 and TYK2 (IC50, 75, 360, 66 nM, respectively).

BMS-911543 is a selective JAK2 inhibitor, with IC50s of 1.1 nM, less selective at JAK1, JAK3 and TYK2 (IC50, 75, 360, 66 nM, respectively).

Specifications

Chemical Name or Material BMS-911543
CAS 1271022-90-2
Color White
Purity Grade Notes Research
Molecular Formula C23H28N8O
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (57.80 mM; Need ultrasonic)
SMILES O=C(C1=CC2=C(N(C)C=N3)C3=C(NC4=NN(C)C(C)=C4)N=C2N1CC)N(C5CC5)C6CC6
Molecular Weight (g/mol) 432.52
Formula Weight 432.52
Percent Purity 97.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.