missing translation for 'onlineSavingsMsg'
Learn More

BMS 833923, TRC

Product Code. 30332831
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30332831

Brand: Toronto Research Chemicals TRCB59632025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material BMS 833923
CAS 1059734-66-5
Molecular Formula C30 H27 N5 O
Quantity 25 mg
Synonym Benzamide, N-[2-methyl-5-[(methylamino)methyl]phenyl]-4-[(4-phenyl-2-quinazolinyl)amino]-, N-[2-Methyl-5-[(methylamino)methyl]phenyl]-4-[(4-phenyl-2-quinazolinyl)amino]benzamide, BMS 833923, BMS-833923, N-[2-Methyl-5-[(methylamino)methyl]phenyl]-4-[(4-phenylquinazolin-2-yl)amino]benzamide
InChI Formula InChI=1S/C30H27N5O/c1-20-12-13-21(19-31-2)18-27(20)33-29(36)23-14-16-24(17-15-23)32-30-34-26-11-7-6-10-25(26)28(35-30)22-8-4-3-5-9-22/h3-18,31H,19H2,1-2H3,(H,33,36)(H,32,34,35)
SMILES CNCc1ccc(C)c(NC(=O)c2ccc(Nc3nc(c4ccccc4)c5ccccc5n3)cc2)c1
IUPAC Name N-[2-methyl-5-(methylaminomethyl)phenyl]-4-[(4-phenylquinazolin-2-yl)amino]benzamide
Molecular Weight (g/mol) 473.57
Formula Weight 473.2216
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.