missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 833923, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | BMS 833923 |
| CAS | 1059734-66-5 |
| Molecular Formula | C30 H27 N5 O |
| Quantity | 100 mg |
| Synonym | Benzamide, N-[2-methyl-5-[(methylamino)methyl]phenyl]-4-[(4-phenyl-2-quinazolinyl)amino]-, N-[2-Methyl-5-[(methylamino)methyl]phenyl]-4-[(4-phenyl-2-quinazolinyl)amino]benzamide, BMS 833923, BMS-833923, N-[2-Methyl-5-[(methylamino)methyl]phenyl]-4-[(4-phenylquinazolin-2-yl)amino]benzamide |
| InChI Formula | InChI=1S/C30H27N5O/c1-20-12-13-21(19-31-2)18-27(20)33-29(36)23-14-16-24(17-15-23)32-30-34-26-11-7-6-10-25(26)28(35-30)22-8-4-3-5-9-22/h3-18,31H,19H2,1-2H3,(H,33,36)(H,32,34,35) |
| SMILES | CNCc1ccc(C)c(NC(=O)c2ccc(Nc3nc(c4ccccc4)c5ccccc5n3)cc2)c1 |
| IUPAC Name | N-[2-methyl-5-(methylaminomethyl)phenyl]-4-[(4-phenylquinazolin-2-yl)amino]benzamide |
| Molecular Weight (g/mol) | 473.57 |
| Formula Weight | 473.2216 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction