missing translation for 'onlineSavingsMsg'
Learn More

BMS-794833, MedChemExpress

Product Code. 16480357
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480357

Brand: Medchem Express HY10497

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-794833 is a VEGFR2 and Met inhibitor extracted from patent WO2009094417, compound example 1; has IC50s of 15 and 1.7 nM, respectively.

BMS-794833 is a VEGFR2 and Met inhibitor extracted from patent WO2009094417, compound example 1

Specifications

Chemical Name or Material BMS-794833
CAS 1174046-72-0
Color White
Purity Grade Notes Research
Molecular Formula C23H15ClF2N4O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (213.29 mM)
SMILES ClC1=C(OC2=CC=C(NC(C3=CNC=C(C4=CC=C(F)C=C4)C3=O)=O)C=C2F)C=CN=C1N
Molecular Weight (g/mol) 468.84
Formula Weight 468.84
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.