missing translation for 'onlineSavingsMsg'
Learn More

GSK-J4, MedChemExpress

Product Code. 16456718
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16456718

Brand: Medchem Express HY15648B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK-J4 is a potent dual inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A with IC50s of 8.6 and 6.6 μM, respectively. GSK-J4 inhibits LPS-induced TNF-α production in human primary macrophages with an IC50 of 9 μM. GSK J4 is a cell permeable prodrug of GSK-J1. GSK-J4 induces endoplasmic reticulum stress-related apoptosis.

GSK-J4 is a potent dual inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A with IC50s of 8.6 and 6.6 μM, respectively. GSK-J4 inhibits LPS-induced TNF-α production in human primary macrophages with an IC50 of 9 μM. GSK J4 is a cell permeable prodrug of GSK-J1. GSK-J4 induces endoplasmic reticulum stress-related apoptosis.

Specifications

Chemical Name or Material GSK-J4
Physical Form Oil
CAS 1373423-53-0
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H27N5O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 36 mg/mL (86.23 mM)
SMILES O=C(OCC)CCNC1=NC(C2=CC=CC=N2)=NC(N3CCC(C=CC=C4)=C4CC3)=C1
Molecular Weight (g/mol) 417.5
Formula Weight 417.5
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.