missing translation for 'onlineSavingsMsg'
Learn More

BMS-754807, MedChemExpress

Product Code. 16496407
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496407 5 mg 5mg
16423678 10 mg 10mg
16420067 50 mg 50mg
16424756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496407 Supplier Medchem Express Supplier No. HY10200
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BMS-754807 is a potent and reversible IGF-1R/IR inhibitor (IC50=1.8 and 1.7 nM, respectively; Ki=IC50 values of 6, 44, 7, 4, 9, and 25 nM, respectively.

BMS-754807 is a potent and reversible IGF-1R/IR inhibitor (IC50=1.8 and 1.7 nM, respectively

Specifications

Chemical Name or Material BMS-754807
CAS 1001350-96-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H24FN9O
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (216.69 mM)
SMILES FC1=CC=C(NC([C@@]2(C)N(C(N=C3NC4=NNC(C5CC5)=C4)=NN6C3=CC=C6)CCC2)=O)C=N1
Molecular Weight (g/mol) 461.49
Formula Weight 461.49
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.