missing translation for 'onlineSavingsMsg'
Learn More

BMS-690514, MedChemExpress

Product Code. 16473828
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473828 10 mg 10mg
16440086 2 mg 2mg
16426557 5 mg 5mg
16420207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473828 Supplier Medchem Express Supplier No. HY10333

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-690514 is a potent and orally active inhibitor of EGFR and VEGFR; has IC50s of 5, 20 and 60 nM for EGFR, HER 2 and HER 4, respectively.

BMS-690514 is a potent and orally active inhibitor of EGFR and VEGFR

Specifications

Chemical Name or Material BMS-690514
CAS 859853-30-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H24N6O2
Quantity 10 mg
Solubility Information DMSO : ≥ 25 mg/mL (67.86 mM)
SMILES O[C@@H]1CN(CC[C@H]1N)CC2=C3C(NC4=CC=CC(OC)=C4)=NC=NN3C=C2
Molecular Weight (g/mol) 368.43
Formula Weight 368.43
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.