missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ BMS 688521
Description
BMS 688521 is a potent inhibitor of the LFA-1/ICAM-1 interaction (IC50 values are 2.5 and 78 nM in HUVEC adhesion and mouse specific adhesion assays respectively). Inhibits eosinophil accumulation in the lungs in a mouse allergic eosinophilic lung inflammation model.
Specifications
Specifications
| Chemical Name or Material | 6-[(5S,9R)-9-(4-Cyanophenyl)-3-(3,5-dichlorophenyl)-1-methyl-2,4-dioxo-1,3,7-triazaspiro[4,4]non-7-yl]-3-pyridinecarboxylic acid |
| CAS | 893397-44-9 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Molecular Formula | C26H19Cl2N5O4 |
| Formula Weight | 536.37 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Vous avez repéré une opportunité d'amélioration ?