missing translation for 'onlineSavingsMsg'
Learn More

BMS-687453, MedChemExpress

Product Code. p-200096184
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480447 50 mg 50mg
16452946 25 mg 25mg
16436797 5 mg 5mg
16415066 100 mg 100mg
16404098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480447 Supplier Medchem Express Supplier No. HY10678

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-687453 is a potent and selective PPARα agonist, with an EC50 and IC50 of 10 nM and 260 nM for human PPARα and 4100 nM and >15000 nM for PPARγ in PPAR-GAL4 transactivation assays.

BMS-687453 is a potent and selective PPARα agonist, with an EC50 and IC50 of 10 nM and 260 nM for human PPARα and 4100 nM and >15000 nM for PPARγ in PPAR-GAL4 transactivation assays.

Specifications

Chemical Name or Material BMS-687453
CAS 1000998-59-3
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClN2O6
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (224.79 mM)
SMILES ClC(C=C1)=CC=C1C2=NC(COC3=CC=CC(CN(C(OC)=O)CC(O)=O)=C3)=C(C)O2
Molecular Weight (g/mol) 444.86
Formula Weight 444.86
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.