missing translation for 'onlineSavingsMsg'
Learn More

BMS-536924, MedChemExpress

Product Code. 16446477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446477 5 mg 5mg
16483748 10 mg 10mg
16420137 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16446477 Supplier Medchem Express Supplier No. HY10262
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BMS-536924 is an orally active, competitive and selective insulin-like growth factor receptor (IGF-1R) kinase and insulin receptor (IR) inhibitor with IC50s of 100 nM and 73 nM, respectively. BMS-536924 has anti-cancer activity.

BMS-536924 is an orally active, competitive and selective insulin-like growth factor receptor (IGF-1R) kinase and insulin receptor (IR) inhibitor with IC50s of 100 nM and 73 nM, respectively. BMS-536924 has anti-cancer activity.

Specifications

Chemical Name or Material BMS-536924
Physical Form Solid
CAS 468740-43-4
Color Brownish Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H26ClN5O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (104.18 mM; Need ultrasonic)
SMILES O=C1C(C2=NC3=CC(N4CCOCC4)=CC(C)=C3N2)=C(C=CN1)NC[C@@H](O)C5=CC=CC(Cl)=C5
Molecular Weight (g/mol) 479.96
Formula Weight 479.96
Percent Purity 95.78%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.