missing translation for 'onlineSavingsMsg'
Learn More

BMS-309403, MedChemExpress

Product Code. 16493628
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493628 10 mg 10mg
16422806 25 mg 25mg
16416367 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493628 Supplier Medchem Express Supplier No. HY101903
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BMS-309403 is a potent, orally active and selective adipocyte fatty acid binding protein (also known as FABP4, aP2) inhibitor with Kis of <2, 250, and 350 nM for FABP4, FABP3, and FABP5, respectively.

BMS-309403 interacts with the fatty-acid-binding pocket within the interior of the protein and competitively inhibits the binding of endogenous fatty acids. BMS-309403 improves endothelial function in apolipoprotein E-deficient mice and in cultured human endothelial cells.

Specifications

Chemical Name or Material BMS-309403
CAS 300657-03-8
Color White
Purity Grade Notes Research
Molecular Formula C31H26N2O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (210.73 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)COC1=CC(C2=CC=CC=C2N3N=C(C4=CC=CC=C4)C(C5=CC=CC=C5)=C3CC)=CC=C1
Molecular Weight (g/mol) 474.55
Formula Weight 474.55
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.