missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS-265246, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
BMS-265246 is a potent and selective cyclin-dependent kinase CDK1 and CDK2 inhibitor, with IC50 values of 6 and 9 nM, respectively. BMS-265246 inhibits CHI3L1 (chitinase 3-like-1) stimulation of ACE2 (angiotensin converting enzyme 2) and SPP (viral spike protein priming proteases). BMS-265246 can be used for ovarian cancer and COVID-19 research.
Specifications
Specifications
| Chemical Name or Material | BMS-265246 |
| CAS | 582315-72-8 |
| Color | Pink |
| Purity Grade Notes | Research |
| Molecular Formula | C18H17F2N3O2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 12.5 mg/mL (36.20 mM; Need ultrasonic) |
| SMILES | CC1=CC(F)=C(C(C2=CN=C(NN=C3)C3=C2OCCCC)=O)C(F)=C1 |
| Molecular Weight (g/mol) | 345.34 |
| Formula Weight | 345.34 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction