missing translation for 'onlineSavingsMsg'
Learn More

BMS-202, MedChemExpress

Product Code. 16487878
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487878 10 mg 10mg
16463827 50 mg 50mg
16437696 100 mg 100mg
16433686 25 mg 25mg
16420348 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487878 Supplier Medchem Express Supplier No. HY19745
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BMS-202 is a potent and nonpeptidic PD-1/PD-L1 complex inhibitor with an IC50 of 18 nM and a KD of 8 μM. BMS-202 binds to PD-L1 and blocks human PD-1/PD-L1 interaction. BMS-202 has antitumor activity.

BMS-202 is a potent and nonpeptidic PD-1/PD-L1 complex inhibitor with an IC50 of 18 nM and a KD of 8 μM. BMS-202 binds to PD-L1 and blocks human PD-1/PD-L1 interaction. BMS-202 has antitumor activity.

Specifications

Chemical Name or Material BMS-202
Physical Form Solid
CAS 1675203-84-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H29N3O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (238.37 mM)
SMILES CC(NCCNCC1=CC=C(OCC2=C(C)C(C3=CC=CC=C3)=CC=C2)N=C1OC)=O
Molecular Weight (g/mol) 419.52
Formula Weight 419.52
Percent Purity 98.38%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.