missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BMS 195614
Selective RARα antagonist
£256.75 - £993.79
Specifications
Purity | >98% |
---|---|
Gene Alias | BMS614 |
Product Type | BMS 195614 |
Molecular Weight (g/mol) | 448.51 |
Inhibitors | BMS 195614 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18015612
|
Tocris Bioscience™
3660/10 |
10 mg |
£256.75
10mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18045202
|
Tocris Bioscience™
3660/50 |
50 mg |
£993.79
50mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Neutral retinoic acid receptor (RAR) α-selective antagonist (Ki = 2.5 nM). Displays no significant effect on nuclear receptor corepressor (NCoR) binding; moderately decreases SMRT binding to RAR. Antagonizes agonist-induced coactivator (CoA) recruitment.Specifications
>98% | |
BMS 195614 | |
BMS 195614 |
BMS614 | |
448.51 |
For Research Use Only