missing translation for 'onlineSavingsMsg'
Learn More

BMS-191011, MedChemExpress

Product Code. 16484208
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484208 10 mg 10mg
16466927 5 mg 5mg
16425166 100 mg 100mg
16420557 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484208 Supplier Medchem Express Supplier No. HY108593
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BMS-191011 (BMS-A) is an opener of the large-conductance, Ca2+-activated potassium (maxi-K) channel, effective in stroke models.

BMS-191011 (BMS-A) is an opener of the large-conductance, Ca2+-activated potassium (maxi-K) channel, effective in stroke models.

Specifications

Chemical Name or Material BMS-191011
CAS 202821-81-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H10ClF3N2O3
Quantity 10 mg
Synonym BMS-A
Solubility Information DMSO : 125 mg/mL (337.19 mM; Need ultrasonic)
SMILES O=C1OC(C2=CC=C(C(F)(F)F)C=C2)=NN1CC3=CC(Cl)=CC=C3O
Molecular Weight (g/mol) 370.71
Formula Weight 370.71
Percent Purity 98.83%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.