missing translation for 'onlineSavingsMsg'
Learn More

BMS-1166, MedChemExpress

Product Code. 16484756
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484756 100 mg 100mg
16470067 50 mg 50mg
16432826 25 mg 25mg
16466417 5 mg 5mg
16473678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16484756 Supplier Medchem Express Supplier No. HY102011
 Restricted Item
Estimated Shipment: 09-10-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress BMS-1166 is a potent PD-1/PD-L1 immune checkpoint inhibitor. BMS-1166 induces dimerization of PD-L1 and blocks its interaction with PD-1, with an IC50 of 1.4 nM. BMS-1166 antagonizes the inhibitory effect of PD-1/PD-L1 immune checkpoint on T cell activation.

BMS-1166 is a potent PD-1/PD-L1 immune checkpoint inhibitor. BMS-1166 induces dimerization of PD-L1 and blocks its interaction with PD-1, with an IC50 of 1.4 nM. BMS-1166 antagonizes the inhibitory effect of PD-1/PD-L1 immune checkpoint on T cell activation.

Especificaciones

Chemical Name or Material BMS-1166
CAS 1818314-88-3
Color White
Purity Grade Notes Research
Molecular Formula C36H33ClN2O7
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (194.97 mM; Need ultrasonic)
SMILES O=C(O)[C@@H]1N(CC2=CC(Cl)=C(OCC3=CC=CC(C4=CC=C(OCCO5)C5=C4)=C3C)C=C2OCC6=CC=CC(C#N)=C6)C[C@H](O)C1
Molecular Weight (g/mol) 641.11
Formula Weight 641.11
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.