Learn More
Invitrogen™ Human BMPR1B (ALK6), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for BMPR1B (ALK6) is NP_001194.1.
Specifications
Specifications
| Accession Number | O00238 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 658 |
| Molecular Weight (g/mol) | 66.7 kDa |
| Name | Human BMPR1B (ALK6), GST Tag |
| Quantity | 10 μg |
| Gene Alias | Activin receptor-like kinase 6; Acvrlk6; AI385617; Alk6; ALK-6; AMDD; AV355320; BDA1D; BDA2; BMP type-1 B receptor; BMP15; Bmpr1b; BMPR-1 B; BMPRIB; BMPR-IB; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; bone morphogenetic protein receptor type 1 B; bone morphogenetic protein receptor type IB; bone morphogenetic protein receptor type-1 B; bone morphogenetic protein receptor, type 1 B; bone morphogenetic protein receptor, type IB; CDw293; CFK-43 A; serine/threonine receptor kinase; serine/threonine-protein kinase receptor R6; SKR6 |
| Common Name | CDw293 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.