missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMPR1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 386000
This item is not returnable.
View return policy
Description
BMPR1A Polyclonal Antibody for Western Blot, ICC/IF, IP
BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases which phosphorylates intracellular SMADs in response to bone morphogenic proteins. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.
Specifications
| BMPR1A | |
| Polyclonal | |
| Unconjugated | |
| BMPR1A | |
| 10q23del; 1110037I22Rik; activin A receptor, type II-like kinase 3; activin receptor-like kinase 3; ACVRLK3; ALK3; ALK-3; AU045487; BMP type-1A receptor; BMP-2/BMP-4 receptor; Bmpr; Bmpr1a; BMPR-1A; BMPRIA; BMPR-IA; Bone morphogenetic protein 4 receptor; bone morphogenetic protein receptor type 1A; bone morphogenetic protein receptor type IA; bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type 1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12166, 657, 81507 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P36894, P36895, Q78EA7 | |
| BMPR1A | |
| Synthetic peptide derived from an internal region of the human BMPR-1A (bone morphogenetic protein receptor type 1A, Serine/threonine protein kinase receptor R5 (SKR5), activin receptor-like kinase-3 (ALK-3)), which differs from mouse and rat BMPR-1A by one nonconservative amino acid replacement. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction