Learn More
Invitrogen™ Human BMPR1A (ALK3), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.
Specifications
Specifications
| Accession Number | P36894 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris with no preservative |
| Gene ID (Entrez) | 657 |
| Molecular Weight (g/mol) | 67 kDa |
| Name | Human BMPR1A (ALK3), GST Tag |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | -80°C, Avoid Freeze/Thaw Cycles |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.