missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR-II Antibody (MM0060-9A10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12064H
This item is not returnable.
View return policy
Description
BMPR-II Monoclonal specifically detects BMPR-II in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BMPR-II | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| BMPR2 | |
| Human recombinant BMPR-II (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Detects human BMPR-II. No cross-reactivity was found to human BMPR-IA and R-IB | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0060-9A10 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| BMP type II receptor, BMP type-2 receptor, BMPR-2, BMPR3, BMPR-IIprimary pulmonary hypertension 1, Bone morphogenetic protein receptor type II, bone morphogenetic protein receptor type-2, bone morphogenetic protein receptor, type II (serine/threonine kinase), BRK-3, EC 2.7.11, EC 2.7.11.30, FLJ41585, FLJ76945, PPH1BMR2, T-ALK, type II activin receptor-like kinase, type II receptor for bone morphogenetic protein-4 | |
| Mouse | |
| Protein G purified | |
| Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
| 659 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction