missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR-IB/ALK-6 Antibody (MM0055-3E12), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12031AF700
This item is not returnable.
View return policy
Description
BMPR-IB/ALK-6 Monoclonal specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BMPR-IB/ALK-6 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BMPR1B | |
| Human recombinant BMPR-IB/ALK-6 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| In direct ELISAs and Western blots, 100% good cross-reactivity with recombinant mouse BMPR-IB, but no cross-reactivity with recombinant human and mouse BMPR-IA is observed. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0055-3E12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR-1B, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase | |
| Mouse | |
| Protein G purified | |
| Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
| 658 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction