missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR-IA/ALK-3 Antibody (MM0053-8L7), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12029AF750
This item is not returnable.
View return policy
Description
BMPR-IA/ALK-3 Monoclonal specifically detects BMPR-IA/ALK-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BMPR-IA/ALK-3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BMPR1A | |
| Human recombinant BMPR-IA/ALK-3 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| In Western blots, this antibody shows good cross-reactivity with mouse BMPR-IA/ALK-3, but no cross-reactivity with recombinant human BMPR-IB or rhBMPR-II. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0053-8L7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ACVRLK310q23del, ALK-3, ALK3EC 2.7.11.30, BMP type-1A receptor, BMPR-1A, bone morphogenetic protein receptor type-1A, bone morphogenetic protein receptor, type IA, CD292, CD292 antigen, EC 2.7.11, Serine/threonine-protein kinase receptor R5, SKR5, type II-like kinase 3 | |
| Mouse | |
| Protein G purified | |
| Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
| 657.0 | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction