missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR-IA/ALK-3 Antibody (002), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90398AF594
This item is not returnable.
View return policy
Description
BMPR-IA/ALK-3 Monoclonal specifically detects BMPR-IA/ALK-3 in Mouse samples. It is validated for ELISA.
Specifications
| BMPR-IA/ALK-3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 2 | |
| ELISA | |
| ACVRLK310q23del, ALK-3, ALK3EC 2.7.11.30, BMP type-1A receptor, BMPR-1A, bone morphogenetic protein receptor type-1A, bone morphogenetic protein receptor, type IA, CD292, CD292 antigen, EC 2.7.11, Serine/threonine-protein kinase receptor R5, SKR5, type II-like kinase 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse BMPR-IA/ALK-3 (NP_033888.2; Met 1-Arg 152). | |
| 0.1 mL | |
| Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
| 657 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction