missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587966
This item is not returnable.
View return policy
Description
Immunogen sequence: SAGAGSHCQK TSLRVNFEDI GWDSWIIAPK EYEAYECKGG CFFPLADDVT PTKHAIVQTL VHLKFPTKVG KACCVPTKLS PISVLYKDDM GVPTLKYHYE GMSVAECGCR; Positive Samples: Mouse pancreas, Mouse skeletal muscle, Rat skeletal muscle.
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons.
Specifications
| BMP9 | |
| Polyclonal | |
| Unconjugated | |
| GDF2 | |
| BMP; BMP9; BMP-9; Bone morphogenetic protein; Bone morphogenetic protein 9; Gdf2; GDF-2; growth differentiation factor 2; growth/differentiation factor 2; HHT5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12165, 290921 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.77 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9WV56 | |
| GDF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-429 of human GDF2 (NP_0572881). | |
| 100 μL | |
| Primary | |
| Mouse, Rat, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction