missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12221-1-AP-20UL
This item is not returnable.
View return policy
Description
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.Specifications
| BMP7 | |
| Polyclonal | |
| Unconjugated | |
| BMP7 | |
| BMP 7, BMP7, bone morphogenetic protein 7, Eptotermin alfa, OP 1, OP1, Osteogenic protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 655 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18075 | |
| BMP7 | |
| BMP7 Fusion Protein Ag2902 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction