missing translation for 'onlineSavingsMsg'
Learn More

BMP6 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

£119.00 - £295.00

Specifications

Antigen BMP6
Concentration 800 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234242
View Documents
Proteintech
55421-1-AP-20UL
20 μL
ONLINE EXCLUSIVE PRICE
£135.00
£119.00 / 20µL
Save £16.00
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236686
View Documents
Proteintech
55421-1-AP-150UL
150 μL
ONLINE EXCLUSIVE PRICE
£349.00
£295.00 / 150µL
Save £54.00
15% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The BMP6 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human BMP6. This antibody recognizes human, mouse antigen. The BMP6 antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications

Specifications

BMP6
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Rabbit
Human, Mouse
Peptide
Primary
Store at -20°C. Stable for one year after shipment.
800 μg/mL
Polyclonal
Liquid
RUO
654
IgG
Antigen affinity purification
Polyclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.