missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP6 Monoclonal Antibody (D2), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£161.00 - £333.00

Specifications

Antigen BMP6
Clone D2
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17900533
View Documents
Invitrogen™
MA551040
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17929492
GREENER_CHOICE
View Documents
Invitrogen™
MA551039
200 μL
£333.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BMP6 is a member of bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, Bmp6 has a proposed role in early development. In addition, the fact that BMP6 is closely related to BMP5 and BMP7 has lead to speculation of possible bone inductive activity.
Specifications

Specifications

BMP6
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
BMP; BMP 6; BMP6; BMP-6; Bone morphogenetic protein; Bone morphogenetic protein 6; D13Wsu115e; vegetal related growth factor (TGFB-related); vegetal-related (TGFB related) cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence; Vgr; Vgr1; VGR-1
BMP6
IgG1 κ
Protein A/G
BMP6
D2
Immunohistochemistry (Paraffin), Western Blot, Western Blot
Unconjugated
Mouse
Human, Pig
P22004
100155536, 654
Recombinant Human BMP6 protein, Ser375-His513 (Accession #P22004).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.