missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578878
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, Mouse Liver Tissue, A549 whole cell. IHC: mouse liver tissue, rat lung tissue, human lung cancer tissue. Flow: U20S cell.
TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth, and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling, and maintenance of a variety of other tissues and organs. BMP-5 is expressed in the nervous system, lungs and liver. It is a known regulator for dendritic growth in sympathetic neurons. BMP-5 is a 454 amino acid precursor protein that is cleaved to release the biologically active C-terminal mature protein.
Specifications
| BMP5 | |
| Polyclonal | |
| Unconjugated | |
| Bmp5 | |
| AU023399; BMP; BMP 5; Bmp5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein; bone morphogenetic protein 5; MGC34244; se; short ear | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12160, 315824, 653 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P22003, P49003 | |
| Bmp5 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human BMP-5 (332-365aa HQDSSRMSSVGDYNTSEQKQACKKHELYVSFRDL). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction