missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bmp4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102424
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BMP4.
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
Specifications
| Bmp4 | |
| Polyclonal | |
| Unconjugated | |
| BMP4 | |
| BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12159, 25296, 652 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12644, P21275, Q06826 | |
| BMP4 | |
| A synthesized peptide derived from human BMP4(Accession P12644), corresponding to amino acid residues L271-G321. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction