missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP4 Mouse anti-Human, Rat, Clone:3C11C7, Abnova™
Mouse monoclonal antibody raised against recombinant human BMP4.
Brand: Abnova MAB17159.100ug
This item is not returnable.
View return policy
Beskrivning
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5′ untranslated region of this gene has been described and three variants are described, all encoding an identical protein. [provided by RefSeq]Specifikationer
| BMP4 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:100-1:500) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| BMP4 | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
| 3C11C7 | |
| bone morphogenetic protein 4 | |
| In PBS (0.05% sodium azide) | |
| BMP2B, BMP2B1, MCOPS6, ZYME | |
| Recombinant protein corresponding to amino acid 277-408 of human BMP4 from E. coli. | |
| 652 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering