missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP3 Monoclonal Antibody (D3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543936
This item is not returnable.
View return policy
Description
BMP3 Monoclonal Antibody for Western Blot, IHC (P)
The BMPs belong to the TGF-Beta superfamilyBMPs stimulate the production of bone matrix proteins and thereby alter stromal cell and osteoclast proliferation, a key step in bone marrow development. In addition to promoting ectopic bone and cartilage development, BMPs regulate the growth, differentiation, chemotaxis, proliferation, and apoptosis of various cell types, including mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells. BMPs also play a role in dorsal/ventral patterning. BMP3, highly expressed in lung, ovary and small intestine, participates in cartilage and bone formation. BMP3 and BMP2 genes map to conserved regions between human and mouse.
Specifications
| BMP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P12645, P22444 | |
| BMP3 | |
| Recombinant protein Bone Morphogenetic Protein 3. | |
| 20 μL | |
| Primary | |
| Human, Bovine, Sheep | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| D3 | |
| Unconjugated | |
| BMP3 | |
| 9130206H07; 9530029I04Rik; BMP; Bmp3; Bmp-3; BMP3A; BMP-3A; Bone morphogenetic protein; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); bone morphogenetic protein 3A; bone morphogenetic protein-3; osteogenin; PBMP3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 101108049, 539527, 651 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction