missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bmp1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103660
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BMP1.
This gene encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.
Specifications
| Bmp1 | |
| Polyclonal | |
| Unconjugated | |
| BMP1 | |
| BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12153, 649, 83470 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13497, P98063 | |
| BMP1 | |
| A synthesized peptide derived from human BMP1(Accession P13497), corresponding to amino acid residues D491-A541. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction