missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582506
This item is not returnable.
View return policy
Description
Immunogen sequence: DSEPLNYKDP CKAAAFLGDI ALDEEDLRAF QVQQAVDLRR HTARKSSIKA AVPGNTSTPS CQSTNGQPQR GACGRWRGRS RSRRAATSRP ERVWPDGVIP FVIGGNFTGS Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.
This gene encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.
Specifications
| BMP1 | |
| Polyclonal | |
| Unconjugated | |
| BMP1 | |
| BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 649 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P13497 | |
| BMP1 | |
| Recombinant protein corresponding to Human BMP1. Recombinant protein control fragment (Product #RP-90828). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction