missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-8B Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149177
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
BMP-8B encodes a secreted ligand of the TGF-beta (transforming growth factor-beta) superfamily of proteins. Ligands of this family bind various TGF-beta receptors leading to recruitment and activation of SMAD family transcription factors that regulate gene expression. The encoded preproprotein is proteolytically processed to generate each subunit of the disulfide-linked homodimer. The encoded protein stimulates thermogenesis in brown adipose tissue. Expression of this gene may be downregulated in pancreatic cancer. BMP-8B may have arose from a gene duplication event and its gene duplicate is also present on chromosome 1.
Specifications
| BMP-8B | |
| Polyclonal | |
| Unconjugated | |
| Bmp8b | |
| BMP; BMP8; BMP-8; Bmp8b; BMP-8B; Bone morphogenetic protein; bone morphogenetic protein 8 (osteogenic protein 2); bone morphogenetic protein 8B; dJ118J21.1 (bone morphogenetic protein 8 (osteogenic protein 2)); MGC131757; OP2; OP-2; Op3; Osteogenic protein 2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 12164, 656 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P34820, P55105 | |
| Bmp8b | |
| E. coli-derived recombinant mouse BMP-8b, Thr261-His399. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction