missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-8b Antibody (13E6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42693H
This item is not returnable.
View return policy
Description
BMP-8b Monoclonal specifically detects BMP-8b in Human samples. It is validated for Western Blot, ELISA.
Specifications
| BMP-8b | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| BMP8B | |
| Recombinant human BMP8b (264-402aa) purified from E. coli | |
| 100 μL | |
| Cytokine Research | |
| 656 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 13000000 | |
| Western Blot, ELISA | |
| BMP-8, bone morphogenetic protein 8b, MGC131757 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction