missing translation for 'onlineSavingsMsg'
Learn More

BMP-8b Antibody (13E6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42693

Product Code. 18161648

  • £358.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA

BMP-8b Monoclonal specifically detects BMP-8b in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BMP-8b
Monoclonal
1 mg/mL
Western Blot 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:100
NP_001711
BMP8B
Recombinant human BMP8b (264-402aa) purified from E. coli
100 μL
Cytokine Research
656
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
13000000
Unconjugated
PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
BMP-8, bone morphogenetic protein 8b, MGC131757
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only