Learn More
Invitrogen™ BMP-7 Mouse ELISA Kit
Sandwich ELISA Kit
£504.00 - £2320.00
Specifications
Accession Number | P23359 |
---|---|
Assay Sensitivity | 4 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455183
|
Invitrogen™
EMBMP7 |
96 Tests |
£504.00
1 set |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17105689
|
Invitrogen™
EMBMP7X5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
The Mouse Bone Morphogenic Protein 7 (Ms BMP-7) ELISA quantitates Ms BMP-7 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms BMP-7. Principle of the method The Mouse BMP-7 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Induces cartilage and bone formation. May be the osteoinductive factor responsible for the phenomenon of epithelial osteogenesis. Plays a role in calcium regulation and bone homeostasis.Specifications
P23359 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Mouse | |
Colorimetric Microplate Reader | |
OP1 | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
4 pg/mL | |
Biotin | |
Cytokines & Receptors | |
This ELISA pair antibody detects mouse and human BMP-7. Other species not determined. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
12162 | |
<12% | |
HRP | |
Development | |
RP23-188A4.1, OP1 | |
1 hr. 20 min. |