missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69126
This item is not returnable.
View return policy
Description
BMP-7 Polyclonal specifically detects BMP-7 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BMP-7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BMP7 | |
| Synthetic peptides corresponding to Bmp7 (bone morphogenetic protein 7). The peptide sequence was selected from the N terminal of Bmp7. Peptide sequence: MVAFFKATEVHLRSIRSTGGKQRSQNRSKTPKNQEALRMASVAENSSSDQ. | |
| Affinity purified | |
| RUO | |
| 655 | |
| Human, Rat, Pig, Bovine, Canine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
| Rabbit | |
| 16 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction