missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-7 Antibody (6E5D12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£442.00
Specifications
| Antigen | BMP-7 |
|---|---|
| Clone | 6E5D12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
BMP-7 Monoclonal specifically detects BMP-7 in Human, Mouse, Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| BMP-7 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Cytokine Research | |
| PBS with 0.05% Sodium Azide | |
| 655 | |
| Purified recombinant fragment of human BMP7 (AA: 239-431) expressed in E. Coli. | |
| Primary | |
| 49.3 kDa |
| 6E5D12 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Virus | |
| BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
| BMP7 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title