missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-7 Antibody (4E7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04259UV
This item is not returnable.
View return policy
Description
BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BMP-7 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BMP7 | |
| Recombinant human BMP7 (293-431 aa) purified from E. coli | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 655 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 40000000 | |
| Western Blot, ELISA | |
| BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human BMP7 mAb, clone 4E7, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BMP7 protein. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction