missing translation for 'onlineSavingsMsg'
Learn More

BMP-7 Antibody (164313) [HRP], Novus Biologicals™

Product Code. 30731923 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731923 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731923 Supplier Novus Biologicals Supplier No. FAB3542H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BMP-7 Monoclonal antibody specifically detects BMP-7 in Human samples. It is validated for Immunohistochemistry,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-7
Applications Immunohistochemistry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 164313
Conjugate HRP
Formulation PBS
Gene Alias BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human BMP-7, Arg292-His431, Accession # P18075
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 655
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.