missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BMP™61 Series Printer Ribbon - R4400 Resin, Red

Product Code. 17545126
Change view
Click to view available options
Color:
Red
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17545126 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17545126 Supplier Brady™ Supplier No. 148132
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

BMP™61 Series ribbon cartridges allow users to change ribbons in seconds.

  • One-color ribbon for printing a single color onto a label
  • R4400 Series Resin Formulation offers excellent resistance to solvents, scratches and high heat when printed on the appropriate label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
  • Automatic label formatting with smart cell technology and easy loading

Specifications

Product Type Printer Ribbon
Color Red
For Use With (Equipment) BMP61 Labeling System
Width (English) 4.31 in.
Alternate Part Number M61-R4410-RD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.