missing translation for 'onlineSavingsMsg'
Learn More

BMP-6 Rabbit anti-Human, Mouse, Rat, Clone: 6N5S9, Novus Biologicals™

Product Code. 18387014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387014 20 μg 20µL
18307281 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18387014 Supplier Novus Biologicals Supplier No. NBP31643020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BMP-6 Monoclonal antibody specifically detects BMP-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-6
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 6N5S9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human BMP-6 (P22004). DGPYDKQPFMVAFFKVSEVHVRTTRSASSRRRQQSRNRSTQSQDVARVSSASDYNSSELKTACRKHELYVSFQDLGWQDWIIAPKGYAANYCDGECSFPLN
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 654
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.