missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-6 Rabbit anti-Human, Mouse, Rat, Clone: 6N5S9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16430-100UL
This item is not returnable.
View return policy
Description
BMP-6 Monoclonal antibody specifically detects BMP-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| BMP-6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| 6N5S9 | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human BMP-6 (P22004). DGPYDKQPFMVAFFKVSEVHVRTTRSASSRRRQQSRNRSTQSQDVARVSSASDYNSSELKTACRKHELYVSFQDLGWQDWIIAPKGYAANYCDGECSFPLN | |
| 100 μg | |
| Cytokine Research | |
| 654 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction