missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57272
This item is not returnable.
View return policy
Description
BMP-6 Polyclonal specifically detects BMP-6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| BMP-6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| BMP6 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FKVSEVHVRTTRSASSRRRQQSRNRSTQSQDVARVSSASDYNSSELKTACRKHELY | |
| 100 μL | |
| Cytokine Research | |
| 654 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction