missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-6 Antibody (74219.11) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB120-10859JF525
This item is not returnable.
View return policy
Description
BMP-6 Monoclonal antibody specifically detects BMP-6 in Human samples. It is validated for ELISA,Western Blot,Neutralization
Specifications
| BMP-6 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 | |
| Recombinant full length protein (Human) expressed in CHO cells. | |
| 0.1 mL | |
| Cytokine Research | |
| 654 | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA, Western Blot, Neutralization | |
| 74219.11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction