missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-4 Recombinant Rabbit Monoclonal Antibody (23GB4795), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | BMP-4 |
|---|---|
| Clone | 23GB4795 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089134
![]() |
Invitrogen™
MA552758 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087505
|
Invitrogen™
MA552759 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-4 Recombinant Monoclonal antibody specifically detects BMP-4 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| BMP-4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P12644 | |
| 652 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4795 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME | |
| A synthesized peptide derived from human BMP4 (355-408AA). | |
| Primary | |
| -20°C | |
| BMP4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title