Learn More
Invitrogen™ BMP-4-Cell Lysates Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHBMP4CLX5
Additional Details : Weight : 0.33820kg
Description
The Human Bone Morphogenic Protein 4 (Hu BMP-4) Cell lysate ELISA quantitates Hu BMP-4 in human cell and tissue lysates. The assay will exclusively recognize both natural and recombinant Hu BMP-4. Principle of the method The Human BMP-4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5′ untranslated region of this gene has been described and three variants are described, all encoding an identical protein.Specifications
P12644 | |
15 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Cell Lysates | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
652 | |
<12% | |
Cell lysis buffer, Standard, Pre-coated 96 well plates, Stop Solution, Biotinylated Detection Antibody, Adhesive Plate Covers, Chromogen, Wash Buffer, Assay Diluent Concentrate, SAV-HRP | |
5 x 96 Tests | |
Angiogenesis | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
8.23 pg/mL to 6000 pg/mL | |
15 pg/mL to 6000 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, BMP-6, BMP-7, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
Human | |
Colorimetric Microplate Reader | |
BMP2B,BMP2B1,MCOPS6,OFC11,ZYME | |
<10% | |
HRP | |
RUO | |
Cell Lysate,20 μ;L | |
BMP2B, BMP2B1, MCOPS6, OFC11, ZYME | |
1 hr. 20 min. |