missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-4 Antibody (19B7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26476-50ul
This item is not returnable.
View return policy
Description
BMP-4 Monoclonal antibody specifically detects BMP-4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry
Specifications
| BMP-4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 19B7 | |
| Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME | |
| A synthesized peptide derived from Human BMP-4 [UniProt P12644] | |
| 50 μL | |
| Biologically Active Proteins, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 652 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction